missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Lysine, Thermo Scientific™
74.67 € - 272.36 €
Specifications
Chemical Name or Material | D-Lysine |
---|---|
Melting Point | 218°C |
CAS | 923-27-3 |
Boiling Point | 311°C |
Color | Beige to White |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17720719
|
Thermo Scientific Acros
469950010 |
1 g |
74.67 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17730719
|
Thermo Scientific Acros
469950050 |
5 g |
272.36 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
D-Lysine | |
923-27-3 | |
Beige to White | |
C6 H14 N2 O2 | |
00008234 | |
(R)-2,6-Diaminohexanoic acid | |
LYSINE | |
146.19 | |
Life science |
218°C | |
311°C | |
Conforms | |
Glass Bottle | |
-12° to -16° | |
≤5 % | |
(20°C, 589 nm) (c=1 aq. soln.) | |
Crystalline powder | |
No special requirements |
Safety and Handling
missing translation for 'shelfLife' : 3 years
missing translation for 'einecsNumber' : 213-091-9
Recommended Storage : Refrigerator +4°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
D-Lysine, Thermo Scientific™