missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Sucrose, ACS reagent, Thermo Scientific Chemicals
D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose.
53.06 € - 623.10 €
Chemical Identifiers
CAS | 57-50-1 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.30 |
MDL Number | MFCD00006626 |
InChI Key | CZMRCDWAGMRECN-PWPRYFECNA-N |
Synonym | sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand |
PubChem CID | 5988 |
ChEBI | CHEBI:17992 |
IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10647421
|
Thermo Scientific Acros
424500010 |
1 kg |
53.06 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10319003
|
Thermo Scientific Acros
424500050 |
5 kg |
167.84 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10733734
|
Thermo Scientific Acros
424500250 |
25 kg |
623.10 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D(+)-Sucrose is a disaccharide carbohydrate
• It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose
Applications
• D(+)-Sucrose can be used for protein and nucleic acid purification
• Its fatty acid esters have laboratory research applications in transdermal systems
• It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
Chemical Identifiers
57-50-1 | |
342.30 | |
CZMRCDWAGMRECN-PWPRYFECNA-N | |
5988 | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
C12H22O11 | |
MFCD00006626 | |
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
CHEBI:17992 | |
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
Specifications
57-50-1 | |
White | |
0.03% max. (105°C) | |
Plastic bottle | |
MFCD00006626 | |
15,9012 | |
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
CZMRCDWAGMRECN-PWPRYFECNA-N | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
5988 | |
342.29 | |
0.005% max | |
Crystals |
190.0°C to 192.0°C | |
0.01% max. | |
Authentic | |
C12H22O11 | |
31,424 | |
+66.3° to +66.8° (25°C, 589nm) (c=26, H2O) | |
Solubility in water: 1970g/L (15°C). Other solubilities: moderately soluble in glycerol and pyridine | |
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O | |
342.30 | |
CHEBI:17992 | |
ACS Reagent | |
0.0008 meq/g max | |
D(+)-Sucrose |
Safety and Handling
missing translation for 'einecsNumber' : 200-334-9
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title