missing translation for 'onlineSavingsMsg'
Learn More
D-Sucrose (Molecular Biology), Fisher BioReagents Product Code.: 10638403

D-Sucrose (Molecular Biology), Fisher BioReagents

Product Code. 10638403
1 kg, Poly Bottle
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
This item is not returnable. View return policy

Product Code. 10638403

Brand: Fisher BioReagents BP2201

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For the preparation of density gradients for DNA ultracentrifugation

D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Alpha Vector Sucrose
Melting Point 190°C
Color White
pH 6.5 to 7.5
Ignition Residue 0.01% max.
Loss on Drying 0.03% max.
Assay Percent Range ≥99.9%
Packaging Poly Bottle
Quantity 1 kg
Merck Index 15, 9012
Specific Rotation +66 to +67° (+ or -)
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 342.29
Percent Purity ≥99.9%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material D-Sucrose
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Sucrose (Molecular Biology), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.