Learn More
Daclatasvir, MedChemExpress
MedChemExpress Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.

76.95 € - 291.60 €
Specifications
Chemical Name or Material | Daclatasvir |
---|---|
Physical Form | Solid |
CAS | 1009119-64-5 |
Color | Light Yellow |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16483988
![]() |
Medchem Express
HY-10466 |
10 mg |
91.80 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16471576
![]() |
Medchem Express
HY-10466 |
500 mg |
291.60 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16470986
![]() |
Medchem Express
HY-10466 |
200 mg |
183.60 €
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16434996
![]() |
Medchem Express
HY-10466 |
100 mg |
137.70 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430347
![]() |
Medchem Express
HY-10466 |
50 mg |
122.85 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16406707
![]() |
Medchem Express
HY-10466 |
5 mg |
76.95 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.Specifications
Daclatasvir | |
1009119-64-5 | |
Research | |
COVID-19-anti-virus | |
DMSO : ≥ 40 mg/mL (54.14 mM) | |
738.88 | |
98.19% |
Solid | |
Light Yellow | |
C40H50N8O6 | |
BMS-790052 EBP 883 | |
O=C([C@@H](NC(OC)=O)C(C)C)N1CCC[C@H]1C2=NC=C(N2)C3=CC=C(C=C3)C4=CC=C(C5=CN=C([C@@H]6CCCN6C([C@@H](NC(OC)=O)C(C)C)=O)N5)C=C4 | |
738.88 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.