Learn More
Daclatasvir, MedChemExpress
MedChemExpress Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Brand: Medchem Express HY-10466
Description
Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.Specifications
Daclatasvir | |
1009119-64-5 | |
50 mg | |
C40H50N8O6 | |
BMS-790052 EBP 883 | |
O=C([C@@H](NC(OC)=O)C(C)C)N1CCC[C@H]1C2=NC=C(N2)C3=CC=C(C=C3)C4=CC=C(C5=CN=C([C@@H]6CCCN6C([C@@H](NC(OC)=O)C(C)C)=O)N5)C=C4 | |
738.88 | |
Research |
Solid | |
Light Yellow | |
Research | |
COVID-19-anti-virus | |
DMSO : ≥ 40 mg/mL (54.14 mM) | |
738.88 | |
98.19% |
Product Suggestions
Customers who viewed this item also viewed