Learn More
Memmert™ DAkkS Factory Calibration for UF30
DAkkS factory calibration for a freely selectable temperature for Memmert Ovens.
Brand: Memmert™ UF30E39696
This item is not returnable.
View return policy
Description
- For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
UF30 Oven | |
DAkkS Factory Calibration |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Memmert™ DAkkS Factory Calibration for UF30 > For Use With: UF30
Spot an opportunity for improvement?Share a Content Correction