Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
Brand: BRAND™ 43308
49.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Calibrated density bottle made with Borosilicate glass
- DIN ISO 3507, type Gay-Lussac
- Calibrated to contain (TC, In)
- Includes individual certificate
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
- The volume in cm³ is specified to a precision of 3 decimal places
Specifications
2Pack | |
10 mL | |
Borosilicate Glass | |
Calibrated Density Bottle |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles > Capacity: 10mL
Spot an opportunity for improvement?Share a Content Correction