Learn More
Davercin, MedChemExpress
MedChemExpress Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
Brand:  Medchem Express HY-100584
Encompass_Preferred
 View more versions of this product
Product Code. 16413098
  • 237.60 € / 10mg
Explore more special offers
Description
Description
Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
Specifications
Specifications
Davercin
White
C38H65NO14
Erythromycin Cyclocarbonate
C[C@]([C@H](OC([C@@H]([C@H]([C@H](C)[C@H]1O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)C)=O)CC)(O4)[C@]([C@H](C([C@@H](C[C@]1(O)C)C)=O)C)([H])OC4=O
759.92
Research
COVID-19-immunoregulation
55224-05-0
Research
10 mg
DMSO : ≥ 50 mg/mL (65.80 mM)
759.92
98.0%
Solid