Learn More
Davercin, MedChemExpress
MedChemExpress Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
POA - 405.00 €
Specifications
Chemical Name or Material | Davercin |
---|---|
CAS | 55224-05-0 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C38H65NO14 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16482616
|
Medchem Express
HY-100584 |
25 mg |
405.00 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16480026
|
Medchem Express
HY-100584 |
2 mg |
POA
2mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16455827
|
Medchem Express
HY-100584 |
5 mg |
153.90 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16413098
|
Medchem Express
HY-100584 |
10 mg |
237.60 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.Specifications
Davercin | |
White | |
C38H65NO14 | |
DMSO : ≥ 50 mg/mL (65.80 mM) | |
759.92 | |
98.0% | |
Solid |
55224-05-0 | |
Research | |
Erythromycin Cyclocarbonate | |
C[C@]([C@H](OC([C@@H]([C@H]([C@H](C)[C@H]1O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)C)=O)CC)(O4)[C@]([C@H](C([C@@H](C[C@]1(O)C)C)=O)C)([H])OC4=O | |
759.92 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.