Learn More
Decitabine, MedChemExpress
MedChemExpress Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Brand:  Medchem Express HY-A0004
Encompass_Preferred
 View more versions of this product
Product Code. 16487976
  • 70.20 € / 100mg
Explore more special offers
Description
Description
Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Specifications
Specifications
Decitabine
White
C8H12N4O4
5-Aza-2'-deoxycytidine 5-AZA-CdR NSC 127716
O=C1N=C(N)N=CN1[C@H](O[C@@H]2CO)C[C@@H]2O
228.21
Research
Cancer-programmed cell death
2353-33-5
Research
100 mg
DMSO : ≥ 50 mg/mL (219.10 mM) ∣H2O : 20 mg/mL (87.64 mM; Need ultrasonic)
228.21
95.0%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Decitabine, MedChemExpress