Learn More
Corning™ DeckWorks™ Reload Systems Starter Kits
Contains one DeckWorks Reload System with standard tips and empty hinged racks
Brand:  Corning™ 4134
 View more versions of this product
Product Code. 11393655
  • 79.78 € / 1 set
Estimated Shipment: 15-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.
  • 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
  • 8 empty racks accompany 1000 μL volume tips.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 1000 μL
Natural
Refill
Starter kit
DeckWorks
Non-sterile
Designed to allow to maximize storage space and help reduce the production and waste of plastic materials
Round
Yes
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ DeckWorks™ Reload Systems Starter Kits > Natural; 100-1000 μL