missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Reload Systems Starter Kits

Contains one DeckWorks Reload System with standard tips and empty hinged racks

75.21 € - 127.13 €

Specifications

Sterility Non-sterile
For Use With (Application) Designed to allow to maximize storage space and help reduce the production and waste of plastic materials
Format Refill
Tip Style Round
Packaging Starter kit
View More Specs

Products 5
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
11373655
View Documents
Corning™
4132
1 to 200 μL Yellow
75.21 €
1 set
In Stock
to see stock.
Add to basket
 
11353655
View Documents
Corning™
4130
0.1 to 10 μL Natural
75.21 €
1 set
Estimated Shipment: 01-09-2025
to see stock.
Add to basket
 
11393655
View Documents
Corning™
4134
100 to 1000 μL Natural
79.78 €
1 set
Estimated Shipment: 01-09-2025
to see stock.
Add to basket
 
11383655
View Documents
Corning™
4133
1 to 300 μL Natural
127.13 €
1 set
Estimated Shipment: 01-09-2025
to see stock.
Add to basket
 
11363655
View Documents
Corning™
4131
1 to 200 μL Natural
75.21 €
1 set
Estimated Shipment: 01-09-2025
to see stock.
Add to basket
 
Description

Description

DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.

  • 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
  • 8 empty racks accompany 1000 μL volume tips.
Specifications

Specifications

Non-sterile
Refill
Starter kit
DeckWorks
Designed to allow to maximize storage space and help reduce the production and waste of plastic materials
Round
Yes
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ DeckWorks™ Reload Systems Starter Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.