Learn More
Ohaus™ Defender™ 2000-D24P Bench Scale
Dependable, accurate, and durable bench scale at an economical price.
626.00 €
Specifications
Battery Life 80 Hours with Rechargeable Battery
Connection Type RS232 (Available as an Accessory)
Construction ABS indicator, 430 grade stainless steel platform with painted steel tubular frame, IP65 aluminum load cell
Display Type Red LED
Includes AC Adapter (Included); Rechargeable Battery (Included)
View More Specs
Products 2
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
15779709
View Documents
Ohaus™
30235942
150 g
626.00 €
Each
15606705
View Documents
Ohaus™
30235947
300 g
626.00 €
Each
Description
Description
  • Sturdy industrial design that can stand up to the rigors of industrial weighing
  • Easy-to-read red LED display can be viewed in dim conditions
  • Versatile power capabilities provide unrestricted operation
Specifications
Specifications
80 Hours with Rechargeable Battery
ABS indicator, 430 grade stainless steel platform with painted steel tubular frame, IP65 aluminum load cell
AC Adapter (Included); Rechargeable Battery (Included)
712.00 x 500.00 mm
712.00 mm
500.00 mm
940.000 mm
AC Adapter (Included), Rechargeable Battery (Included)
RS232 (Available as an Accessory)
Red LED
ABS indicator, 430 grade stainless steel platform with painted steel tubular frame, IP65 aluminum load cell
712.00 x 500.00 x 940.000 mm
600 mm
500 mm
712.00 mm