Learn More
OHAUS™ Defender™ 5000 R Series Base
OHAUS™ Defender™ 5000 R Series Base offers durability for demanding industrial applications. The R series features moisture-protected single-point load cells, powder-coated steel construction, non-slip rubber adjustable feet, removable stainless steel weighing pans and leveling bubbles.
503.00 € - 1650.00 €
Specifications
Construction | 304-grade stainless steel platform, sand grey powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet, level indicator |
---|---|
Protection Class | IP67 |
Temperature (Metric) Operating | -10°C to +40°C |
Product Code | Brand | Capacity | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Model | Price | Quantity & Availability | ||||
15908435
|
OHAUS™
30419624 |
300.00 kg, 600.00 kg | D600RTDV |
1650.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928415
|
OHAUS™
30419596 |
6.00 kg, 15.00 kg | D15RQDL |
662.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928425
|
OHAUS™
30419616 |
30.00 kg, 60.00 kg | D60RQDV |
1180.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938415
|
OHAUS™
30419597 |
15.00 kg, 30.00 kg | D30RQDL |
662.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938425
|
OHAUS™
30419617 |
60.00 kg, 150.00 kg | D150RQDV |
1180.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948415
|
OHAUS™
30419608 |
30.00 kg, 60.00 kg | D60RQDL |
662.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948425
|
OHAUS™
30419618 |
150.00 kg, 300.00 kg | D300RQDV |
1180.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988405
|
OHAUS™
30419592 |
3.00 kg, 6.00 kg | D6RQDR |
503.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988415
|
OHAUS™
30419612 |
30.00 kg, 60.00 kg | D60RQDX |
831.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988425
|
OHAUS™
30419622 |
60.00 kg, 150.00 kg | D150RTDV |
1650.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998405
|
OHAUS™
30419593 |
6.00 kg, 15.00 kg | D15RQDR |
503.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998415
|
OHAUS™
30419613 |
60.00 kg, 150.00 kg | D150RQDX |
831.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998425
|
OHAUS™
30419623 |
150.00 kg, 300.00 kg | D300RTDV |
1650.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The R Series features a stainless steel platform, powder-coated steel frame and an aluminum load cell to withstand harsh industrial environments.
- The Defender 5000 bases are equipped with an EasyConnect cable, which allows instant configuration with Defender 5000 (TD52) indicators, speeding up installation.
- Defender 5000 bases are NTEP-certified and Measurement Canada approved.
- These bases also carry OIML and EC Type approvals, which make them ideal for legal-for-trade use.
Specifications
304-grade stainless steel platform, sand grey powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet, level indicator | |
IP67 | |
-10°C to +40°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
OHAUS™ Defender™ 5000 R Series Base