Learn More
Demeclocycline hydrochloride, MedChemExpress
MedChemExpress Demeclocycline hydrochloride is a tetracycline antibiotic; is an antibiotic in the treatment of Lyme disease, acne, and bronchitis.
Brand: Medchem Express HY-17560
This item is not returnable.
View return policy
Description
Demeclocycline hydrochloride is a tetracycline antibioticSpecifications
Demeclocycline hydrochloride | |
Light Yellow | |
C21H22Cl2N2O8 | |
DMSO : 100 mg/mL (199.48 mM; Need ultrasonic) ∣H2O : 20 mg/mL (39.90 mM; Need ultrasonic and warming) | |
501.31 | |
95.09% | |
Powder |
64-73-3 | |
Research | |
500 mg | |
O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@H](O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4Cl)N.Cl | |
501.31 | |
Research | |
COVID-19-immunoregulation |