missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic acid, 99%, Thermo Scientific Chemicals

37.89 € - 260.00 €

Chemical Identifiers

CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400551
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
B20061.14
25 g
37.89 €
25g
Please to purchase this item. Need a web account? Register with us today!
11410551
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
B20061.22
100 g
87.33 €
100g
Please to purchase this item. Need a web account? Register with us today!
11420551
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
B20061.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Deoxycholic acid has been used in a modified procedure to recover 40-80% of a protein from a 1 μg/mL solution. It forms complexes with fatty acid. Used as an emulsifying agent in food, a precursor in the synthesis of cortisone, and a gallbladder stimulant. It has been used to study assess how physiological concentrations of ursodeoxycholic acid (UDCA) vs. deoxycholic acid (DCA) affect barrier function in mouse intestinal tissue. Deoxycholic acid has been used in a study to assess a pH-Responsive Mechanism of a Deoxycholic Acid and Folate Co-Modified Chitosan Micelle under Cancerous Environment. It has also been used in a study to investigate dose-dependent anti-inflammatory effect of ursodeoxycholic acid in experimental colitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxycholic acid has been used in a modified procedure to recover 40-80% of a protein from a 1 µg/mL solution. It forms complexes with fatty acid. Used as an emulsifying agent in food, a precursor in the synthesis of cortisone, and a gallbladder stimulant. It has been used to study assess how physiological concentrations of ursodeoxycholic acid (UDCA) vs. deoxycholic acid (DCA) affect barrier function in mouse intestinal tissue. Deoxycholic acid has been used in a study to assess a pH-Responsive Mechanism of a Deoxycholic Acid and Folate Co-Modified Chitosan Micelle under Cancerous Environment. It has also been used in a study to investigate dose-dependent anti-inflammatory effect of ursodeoxycholic acid in experimental colitis.

Solubility
The free acid is soluble at 0.044 g/L in water at 20°C. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

83-44-3
392.58
KXGVEGMKQFWNSR-LLQZFEROSA-N
222528
(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
C24H40O4
MFCD00003673
deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
CHEBI:28834
[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

83-44-3
C24H40O4
3219882
deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
KXGVEGMKQFWNSR-LLQZFEROSA-N
[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
392.58
CHEBI:28834
99%
172°C to 176°C
MFCD00003673
14,2899
The free acid is soluble at 0.044g/L in water at 20°C. Solubility in methanol is almost transparent.
+54° (c=2 in Ethanol)
(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
222528
392.58
Deoxycholic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 201-478-5

missing translation for 'rtecsNumber' : FZ2100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only