Learn More
Macherey-Nagel™ Developing Chamber
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Brand:  Macherey-Nagel™ 814021
Product Code. 12760213
Description
Description
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Specifications
Specifications
4 Pk.
Developing Chamber
For TLC Micro-Sets
TLC Micro-Sets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Developing Chamber > For Use With: For TLC Micro-Sets