missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Dewax and HIER Buffer L,M,H

Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.

Brand:  Epredia™ TA-999-DHBL

528.00 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 15210715

  • 615.00 € / 1 L
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.

Specifications

Specifications

10 x 100 mL
Dewax and HIER Buffer
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Carbol Fuchsin-Kinyoun Solution (Cold Technique)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray, or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, phenol, basic fuchsin, isopropanol, methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.