missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Dewax and HIER Buffer L,M,H

Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.

615.00 €

Specifications

Product Type Dewax and HIER Buffer
Quantity 10 x 100 mL
View More Specs

Products 3
Product Code Brand Solution Type Price Quantity & Availability  
Product Code Brand Solution Type Price Quantity & Availability  
15200715
View Documents
Epredia™
TA-999-DHBH
Buffer - High
615.00 €
1 L
In Stock
to see stock.
Add to basket
 
15210715
View Documents
Epredia™
TA-999-DHBL
Buffer - Low
615.00 €
1 L
In Stock
to see stock.
Add to basket
 
15220715
View Documents
Epredia™
TA-999-DHBM
Buffer - Medium
615.00 €
1 L
In Stock
to see stock.
Add to basket
 
Description

Description

Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.

Specifications

Specifications

Dewax and HIER Buffer
10 x 100 mL
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : titon-X100, EDTA, tetrasodium EDTA, tris(hydroxymethyl) aminomehtane, 2-methyl-3-isothiazolone, tet

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.