Learn More
Dexamethasone 9,11-epoxide, MedChemExpress
MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Brand:  Medchem Express HY-N0348
Encompass_Preferred
 View more versions of this product
Product Code. 16422376
  • 414.45 € / 500mg
Explore more special offers
Description
Description
Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Specifications
Specifications
Dexamethasone 9,11-epoxide
White
C22H28O5
DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
372.45
99.3%
Solid
24916-90-3
Research
500 mg
C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O
372.45
Research
COVID-19-immunoregulation