Learn More
Dexamethasone 9,11-epoxide, MedChemExpress
MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.

83.70 € - 434.70 €
Specifications
Chemical Name or Material | Dexamethasone 9,11-epoxide |
---|---|
CAS | 24916-90-3 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C22H28O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498948
![]() |
Medchem Express
HY-N0348 |
10 mg |
83.70 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16489056
![]() |
Medchem Express
HY-N0348 |
100 mg |
201.15 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16464817
![]() |
Medchem Express
HY-N0348 |
50 mg |
133.65 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16441506
![]() |
Medchem Express
HY-N0348 |
200 mg |
267.30 €
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16422376
![]() |
Medchem Express
HY-N0348 |
500 mg |
434.70 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.Specifications
Dexamethasone 9,11-epoxide | |
White | |
C22H28O5 | |
C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O | |
372.45 | |
Research | |
COVID-19-immunoregulation |
24916-90-3 | |
Research | |
DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
372.45 | |
99.3% | |
Solid |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.