Learn More
Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals
52.40 € - 855.00 €
Chemical Identifiers
CAS | 68959-06-8 |
---|---|
Molecular Formula | C14H33AlO6Si |
Molecular Weight (g/mol) | 352.48 |
MDL Number | MFCD00053771 |
InChI Key | MTHHTRVJDXYJHE-UHFFFAOYNA-N |
Synonym | di-sec-butoxyaluminoxytriethoxysilane, bis butan-2-olato triethyl orthosilicato-o' aluminum, aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o', aluminum, bis 2-butanolato triethyl orthosilicato-kappao', di-s-butoxyaluminoxy-triethoxysilane, di-sec-butoxy triethoxysilyl oxy aluminum, bis sec-butoxy alumanyl triethyl silicate, aluminum,bis 2-butanolato triethyl orthosilicato-ko', diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane |
PubChem CID | 16684138 |
IUPAC Name | di(butan-2-yloxy)alumanyl triethyl silicate |
SMILES | CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309587
|
Thermo Scientific Alfa Aesar
041210.09 |
10 g |
52.40 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399577
|
Thermo Scientific Alfa Aesar
041210.18 |
50 g |
223.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389577
|
Thermo Scientific Alfa Aesar
041210.30 |
250 g |
855.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68959-06-8 | |
352.48 | |
MTHHTRVJDXYJHE-UHFFFAOYNA-N | |
16684138 | |
CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC |
C14H33AlO6Si | |
MFCD00053771 | |
di-sec-butoxyaluminoxytriethoxysilane, bis butan-2-olato triethyl orthosilicato-o' aluminum, aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o', aluminum, bis 2-butanolato triethyl orthosilicato-kappao', di-s-butoxyaluminoxy-triethoxysilane, di-sec-butoxy triethoxysilyl oxy aluminum, bis sec-butoxy alumanyl triethyl silicate, aluminum,bis 2-butanolato triethyl orthosilicato-ko', diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane | |
di(butan-2-yloxy)alumanyl triethyl silicate |
Specifications
>200°C | |
Colorless | |
C14H33AlO6Si | |
UN1993 | |
di-sec-butoxyaluminoxytriethoxysilane, bis butan-2-olato triethyl orthosilicato-o' aluminum, aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o', aluminum, bis 2-butanolato triethyl orthosilicato-kappao', di-s-butoxyaluminoxy-triethoxysilane, di-sec-butoxy triethoxysilyl oxy aluminum, bis sec-butoxy alumanyl triethyl silicate, aluminum,bis 2-butanolato triethyl orthosilicato-ko', diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane | |
MTHHTRVJDXYJHE-UHFFFAOYNA-N | |
di(butan-2-yloxy)alumanyl triethyl silicate | |
16684138 | |
Moisture sensitive | |
Di-sec-butoxyaluminoxytriethoxysilane |
68959-06-8 | |
Liquid | |
MFCD00053771 | |
46°C (115°F) | |
Hydrolyzes with water. | |
CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC | |
352.48 | |
352.48 | |
1.00 g/mL |
Safety and Handling
GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 273-392-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only