Learn More
MP Biomedicals Diagnostic Kit for 25-Hydroxyvitamin D (25-OH-VD) (MPQuanti™ only)
Diagnostic Kit for 25-Hydroxyvitamin D (25-OH-VD) is intended for in-vitro quantitative determination of total Vitamin D (D2+D3) in serum or plasma.
Brand:  MP Biomedicals 07OHVD5025
Product Code. 30280619
  • 97.97 € / 1 set
Explore more special offers
Description
Description
  • Diagnostic Kit for 25-Hydroxyvitamin D (25-OH-VD) (Serum/Plasma) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad.
  • Vitamin D in the sample will compete with the VD-BSA antigen coated on the membrane.
  • The less Vitamin D in the sample, the more fluorescent microspheres conjugated with anti-VD antibodies can be captured by the VD-BSA antigen coated on the membrane (Test line).
  • The concentration of Vitamin D in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of Vitamin D in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer to show Vitamin D concentration in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 5 -100 ng/mL, (R2) is 0.956
  • Test results should be interpreted at 15 minutes
  • Used as an aid to assessment of Vitamin D levels
Specifications
Specifications
25-OH-Vitamin D
Human serum, Human plasma
Ready-to-Use (RTU)
Immunofluorescence
POC, Quantitative, vitamin D, 25-OH-VD
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for 25-Hydroxyvitamin D (25-OH-VD) (MPQuanti™ only) > 25-Hydroxyvitamin D