Learn More
MP Biomedicals Diagnostic Kit for CK-MB (MPQuanti™ only)
Diagnostic Kit for CK-MB is a chromatographic immunoassay for the quantitative detection of human CK-MB in human whole blood, serum or plasma.
Brand:  MP Biomedicals 07CKMB5025
Product Code. 30280615
  • 65.32 € / 1 set
Explore more special offers
Description
Description
  • The CK-MB Test Cassette (Whole Blood/Serum/Plasma) is a chromatographic immunoassay for the quantitative detection of human CK-MB in human whole blood, serum or plasma as an aid in the diagnosis of myocardial infarction (MI).
  • The sample moves through the strip from sample pad to absorbent pad.
  • If the test sample contains CK-MB, it attaches to the CK-MB antibody which is conjugated with fluorescent microspheres.
  • Then the complex will be captured by the capture antibody coated on the nitrocellulose membrane (Test line).
  • The concentration of CK-MB in the sample correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and product standard curve, the concentration of CK-MB in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE-marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 0. 2 ̴75ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
Specifications
Specifications
CK-MB
Human whole blood, Human serum, Human plasma
Ready-to-Use (RTU)
Immunofluorescence
POC, CK-MB, Quantitative
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for CK-MB (MPQuanti™ only) > CK-MB