Learn More
Diamond powder, synthetic, <1 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Diamond powder is generally used as an abrasive to grind and polish hard surfaces
Brand:  Thermo Scientific Alfa Aesar 013401.06
 View more versions of this product
Product Code. 11356718
  • 203.01 € / 5g
Explore more special offers
Description
Description
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Diamond powder
Powder
(metals basis)
99.9%
Synthetic
5 g
MFCD00211867