missing translation for 'onlineSavingsMsg'
Learn More

Diamond powder, synthetic, <1 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Diamond powder is generally used as an abrasive to grind and polish hard surfaces

71.86 € - 203.01 €

Specifications

Chemical Name or Material Diamond powder
Name Note Synthetic
Physical Form Powder
Assay Percent Notes (metals basis)
MDL Number MFCD00211867
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346718
View Documents
Thermo Scientific Alfa Aesar
013401.03
1 g
71.86 €
1g
Please to purchase this item. Need a web account? Register with us today!
11356718
View Documents
Thermo Scientific Alfa Aesar
013401.06
5 g
203.01 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diamond powder
Powder
MFCD00211867
Synthetic
(metals basis)
99.9%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-953-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.