Learn More
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C
22.41 € - 166.83 €
Chemical Identifiers
CAS | 75-09-2, 513-35-9 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10487532
|
Thermo Scientific Acros
348461000 |
100 mL | AcroSeal™ Glass Bottle |
22.41 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10354263
|
Thermo Scientific Acros
348465000 |
500 mL | AcroSeal™ Glass Bottle |
55.98 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10387841
|
Thermo Scientific Acros
348460010 |
1 L | AcroSeal™ Glass Bottle |
85.73 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295661
|
Thermo Scientific Acros
348460025 |
2.5 L | AcroSeal™ Glass Bottle |
166.83 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-09-2, 513-35-9 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
75-09-2, 513-35-9 | |
-97.0°C | |
1.3250g/mL | |
Authentic | |
CH2Cl2 | |
MFCD00000881 | |
01,676; 02,273; 04,337; 07,239; 13,106 | |
1.325 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
ClCCl | |
50ppm (approx.) amylene | |
6344 | |
0.43 mPa.s (20°C) | |
475hPa at 20°C | |
Extra Dry over Molecular Sieve | |
Dichloromethane |
>120°C | |
Colorless | |
39.0°C to 40.0°C | |
99.8% min. (GC) | |
CH2Cl2 | |
01, 60 | |
15, 6135 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
84.93 | |
CHEBI:15767 | |
84.93 | |
99.8% | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause drowsiness or dizziness.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-838-9
- Dichloromethane
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Dichloromethane
RUO – Research Use Only