Learn More
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals
69.20 € - 110.00 €
Chemical Identifiers
CAS | 75-09-2, 67-56-1 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10180272
|
Thermo Scientific Acros
364230010 |
1 L | Glass Bottle |
69.20 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10190272
|
Thermo Scientific Acros
364230025 |
2.5 L | Glass Bottle |
110.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLCChemical Identifiers
75-09-2, 67-56-1 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-97.0°C | |
75-09-2, 67-56-1 | |
1.3250g/mL | |
Liquid | |
0.0002% max. | |
CH2Cl2 | |
MFCD00000881 | |
01, 60 | |
01,676; 02,273; 04,337; 07,239; 13,106 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
HPLC |
(1 cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.02 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm | |
0.0001 meq/g max. | |
39.0°C to 40.0°C | |
99.80% | |
1.325 | |
CH2Cl2 | |
0.43 mPa.s (20°C) | |
0.02% max. | |
15, 6135 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
99.8% | |
Dichloromethane, Stabilized |
Safety and Handling
GHS P Statement
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 200-838-9
missing translation for 'rtecsNumber' : PA8050000
missing translation for 'tsca' : TSCA
- Dichloromethane, stabilized with methanol
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only