Learn More
Dicyclohexano-18-crown-6, mixture of isomers, 97%, Thermo Scientific Chemicals
30.85 € - 327.63 €
Chemical Identifiers
CAS | 16069-36-6 |
---|---|
Molecular Formula | C20H36O6 |
Molecular Weight (g/mol) | 372.502 |
MDL Number | MFCD00005099 |
InChI Key | BBGKDYHZQOSNMU-UHFFFAOYSA-N |
Synonym | dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612 |
PubChem CID | 85955 |
SMILES | C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488256
|
Thermo Scientific Alfa Aesar
A15344.03 |
1 g |
30.85 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374509
|
Thermo Scientific Alfa Aesar
A15344.09 |
10 g |
169.85 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498256
|
Thermo Scientific Alfa Aesar
A15344.14 |
25 g |
327.63 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.
Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
Chemical Identifiers
16069-36-6 | |
372.502 | |
BBGKDYHZQOSNMU-UHFFFAOYSA-N | |
85955 |
C20H36O6 | |
MFCD00005099 | |
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612 | |
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2 |
Specifications
16069-36-6 | |
>110°C (230°F) | |
C20H36O6 | |
UN2811 | |
Hygroscopic | |
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612 | |
BBGKDYHZQOSNMU-UHFFFAOYSA-N | |
372.502 | |
372.5 | |
Dicyclohexano-18-crown-6, mixture of isomers |
46°C to 53°C | |
Odorless | |
MFCD00005099 | |
1130529 | |
14,2602 | |
Soluble in Toluene. Slightly soluble (13g/L,26°C) in water. | |
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2 | |
85955 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 240-216-4
missing translation for 'rtecsNumber' : HP5385000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.