missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

55.00 € - 212.00 €

Chemical Identifiers

CAS 81-21-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00077209
InChI Key BQQUFAMSJAKLNB-UHFFFAOYSA-N
Synonym dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
PubChem CID 6673
SMILES C1C2C3CC4C(C3C1C5C2O5)O4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347905
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L02940.06
5 g
55.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11357905
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L02940.14
25 g
212.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

81-21-0
164.204
BQQUFAMSJAKLNB-UHFFFAOYSA-N
6673
C10H12O2
MFCD00077209
dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
C1C2C3CC4C(C3C1C5C2O5)O4

Specifications

81-21-0
1.331
Odorless
MFCD00077209
112743
Soluble in water.
C1C2C3CC4C(C3C1C5C2O5)O4
6673
99%
185°C to 189°C
326°C
C10H12O2
UN2811
dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
BQQUFAMSJAKLNB-UHFFFAOYSA-N
164.204
164.2
Dicyclopentadiene diepoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c

H301-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 201-334-1

missing translation for 'rtecsNumber' : PB9625200

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only