missing translation for 'onlineSavingsMsg'
Learn More
Diethyl carbonate, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals Product Code.: 11915161

Diethyl carbonate, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11915161
1 L
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy

Product Code. 11915161

Brand: Thermo Scientific Acros 436130010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-58-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00009107
InChI Key OIFBSDVPJOWBCH-UHFFFAOYSA-N
Synonym ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2coShow MoreShow Less
PubChem CID 7766
IUPAC Name diethyl carbonate
SMILES CCOC(=O)OCC

Specifications

Melting Point -43.0°C
Density 0.9750g/mL
Boiling Point 126.0°C to 128.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5O)2CO
Refractive Index 1.3830 to 1.3850
Quantity 1 L
Merck Index 15,3835
Specific Gravity 0.975
Solubility Information Solubility in water: 18.8g/L.
Viscosity 0.827 mPa.s (20°C)
Formula Weight 118.13
Percent Purity 99%
Chemical Name or Material Diethyl carbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466048 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number436130010,436131000
Lot NumberA0450125 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number436130010,436131000
Lot NumberA0444322 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog Number436130010,436131000
Lot NumberA0434718 Certificate TypeCertificate of Analysis Date28/09/2021 Catalog Number436130010,436131000
Lot NumberA0417718 Certificate TypeCertificate of Analysis Date20/01/2021 Catalog Number436130010,436131000
compliance-icons
Product Identifier
  • Diethyl carbonate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl carbonate, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.