missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Spécification
Color | Brown to Yellow |
Density | 1.3160g/mL |
Boiling Point | 30.0°C to 32.0°C (3.0 mmHg) |
Flash Point | 23°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Plastic bottle |
Linear Formula | (C2H5)2NSF3 |
Refractive Index | 1.4100 to 1.4160 |
Quantity | 5 g |
Afficher plus |
Suggestions de produits
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0439038 | Certificate of Analysis | 15/02/2024 | 216111000,216110250,216110050 |
A0445827 | Certificate of Analysis | 21/09/2023 | 216111000,216110250,216110050 |
A0393621 | Certificate of Analysis | 15/12/2022 | 216111000,216110250,216110050 |
A0413061 | Certificate of Analysis | 13/12/2022 | 216111000,216110250,216110050 |
A0405027 | Certificate of Analysis | 12/12/2022 | 216111000,216110250,216110050 |
Sécurité et traitement


Identifiant produit
- Diethylaminosulfur trifluoride
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Self-reactive substance or mixture Type B
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H241-Heating may cause a fire or explosion.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu