Description
Salmonella poly antiserum
- Febrile control antisera
- Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
- Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
- Used along with febrile antigens as positive or negative controls
- For identification of morphologically and biochemically identified Shigella species
- Can be used only in slide agglutination test
- Lyophilized, polyclonal rabbit antisera
Specifications
For in vitro diagnostic use | |
Shigella | |
3 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Vial Shigella Antis Poly D Lyophilize
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Azide
Product Content Correction
Product Title
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction