missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Poly Antisera

For agglutination test

372.00 € - 795.00 €

Specifications

Certifications/Compliance For in vitro diagnostic use
Product Type Shigella
Quantity 3 mL
View More Specs

Products 4
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11702874
View Documents
BD
227761
Shigella
393.00 €
3mL
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
17352361
View Documents
BD
228351
Shigella
372.00 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
11894422
View Documents
BD
228341
Shigella
795.00 €
5mL
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
11772894
View Documents
BD
228371
Shigella
395.00 €
3mL
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
Description

Description

Salmonella poly antiserum

  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera

Specifications

Specifications

For in vitro diagnostic use
Shigella
3 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vial Shigella Antis Poly A1 Lyophilized

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide

SDS
Documents

Documents

Product Certifications

For in vitro diagnostic use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.