Learn More
Digi-Sense™ Advanced Precalibrated Thermistor Thermometers
Perfect for the lab to production floor
Brand:  Digi-Sense™ 2025093
140.09 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
 View more versions of this product
Product Code. 15651774
  • 160.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Includes: Protective rubber armor boot with built-in stand, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging models 20250-92, -94, and -96 also include a USB cable and software.
Description
Description
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super lager LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
Services
2 Years
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Digi-Sense™ Advanced Precalibrated Thermistor Thermometers > Single input, not data logging