Learn More
Cole-Parmer™ Digi-Sense™ FEP-Insulated Type-K Probes with FEP-Coated Junction
Use these probes for applications requiring an isolated junction. Cole-Parmer™ Digi-Sense™ FEP-Insulated Type-K Probes with FEP-Coated Junction are ideal for long term immersion in semi caustic environments.
Brand:  Cole-Parmer™ 08466-82
Product Code. 11751349
  • 41.90 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Due to the flexibility along with the sealed tip these probes can be used in harsh environments, submerged in liquid or great for general purpose everyday use. Type “K” is the most popular calibration thermocouple. It has the widest temperature range of all calibrations. Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue and type E-purple. Response Time: 2 seconds or greater.
  • Easily cleanable for multiple uses in different applications
  • Made from 24AWG cable for a fast response time along with the ability applied in hard to reach locations
  • Insulated wire probe single wire FEP with FEP over jacket
Specifications
Specifications
-150°C to +204°C
FEP
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Digi-Sense™ FEP-Insulated Type-K Probes with FEP-Coated Junction > Diameter: 2.4mm; Temperature Range: -150 to 204deg.C