Learn More
3M™ Particulate Respirator, Series C100
Provide comfortable, effective respiratory protection against dust and mist. The 3M C100 Series Particulate Respirator is lightweight and comfortable.
Brand:  3M™ 7000088843
9.15 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 13517010
  • 24.55 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.
  • The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
  • Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FFP2 NR D
Polypropylene
Valved Particulate Respirator
C112