missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Digital Calipers

Manufactured from stainless steel. BOCHEM™ Digital Calipers have a high degree of accuracy.

Brand:  Bochem™ 12116

Product Code. 10711961

  • 154.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 5 digits LCD display
  • Accurary +/- 0.03mm
  • Resolution 0.01mm
  • Conversion mm/inch
Specifications

Specifications

150 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Digital Calipers > Length: 150mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.