missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Stainless Steel Sensor for EKT Hei-Con Magnetic Stirrer

Stainless Steel Sensor for use with EKT Hei-Con magnetic stirrer.

Brand:  Heidolph Instruments 509-88000-00

Product Code. 10019070

  • 523.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with EKT Hei-Con magnetic stirrer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 to 1400 rpm
0.1 in.
8.3 in.
Hei-PLATE Mix 'n' Heat Core
EKT Hei-Con
Contact Thermometer Stirrer
Digital
3.5 mm
210 mm
Cable and Stainless Steel temperature sensor
-80°C to +350°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph Instruments Stainless Steel Sensor for EKT Hei-Con Magnetic Stirrer > Stainless steel Sensor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.