Learn More
Bellingham Stanley™ Digital Refractometer RFM340-M
The Bellingham Stanley™ RFM340-M digital refractometer with three-decimal/0-100 Brix (1.58 RI) scales and Peltier control. Perfect for measurement and control in demanding food, beverage, chemical and industrial applications
Brand:  Bellingham Stanley™ 19-45
Product Code. 16339693
  • 14125.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Benchtop model for demanding food, beverage, chemical and industrial Brix or RI applications
  • Tactile and robust keypad with large 10 cm high resolution screen display
  • One-touch configuration of scale, temperature and data storage
  • User-defined methods or selected from the on-board library
  • Flat prism surface for easy cleaning
  • Wide beam scan for non-homogenous samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Benchtop Refractometer
1.32 to 1.58
0.000001 RI, 0.01/0.001 Brix
USB, Ethernet, RS232
100/240 V 50/60 Hz
0 to 100 Brix
±0.00002 (1.32 to 1.38 RI), ±0.00004 (1.38 to 1.58 RI); ±0.010 (0 to 30°Brix), ±0.030 (30 to 100°Brix)
4 sec.
0°C or 10°C below ambient whichever is greater to 70°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bellingham Stanley™ Digital Refractometer RFM340-M > 100 to 240 V, 50/60 Hz