Learn More
Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Thermo Scientific Chemicals
121.61 € - 1422.08 €
Chemical Identifiers
CAS | 12033-81-7 |
---|---|
Molecular Formula | H2N2O8PtS |
Molecular Weight (g/mol) | 385.16 |
MDL Number | MFCD00064668 |
InChI Key | SPPCQWREVYSZTJ-UHFFFAOYSA-N |
Synonym | dihydrogen dinitrosulfatoplatinate ii, dinitroplatinum; sulfuric acid, dihydrogen dinitrosulfatoplatinate ii solution, pt cont. pt |
PubChem CID | 102602532 |
IUPAC Name | platinum;sulfuric acid;dinitrite |
SMILES | [H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450300
|
Thermo Scientific Alfa Aesar
012627.NJ |
∼0.5 g |
121.61 €
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430300
|
Thermo Scientific Alfa Aesar
012627.NA |
∼2 g |
309.54 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440300
|
Thermo Scientific Alfa Aesar
012627.NC |
∼10 g |
1422.08 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dihydrogen dinitrosulfatoplatinate(II) solution is used in the preparation of electrodeposition of platinum films and biocompatible platinum thin film electrodeposition on tin substrates, which acts as the interconnects material for implantable medical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12033-81-7 | |
385.16 | |
SPPCQWREVYSZTJ-UHFFFAOYSA-N | |
102602532 | |
[H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O |
H2N2O8PtS | |
MFCD00064668 | |
dihydrogen dinitrosulfatoplatinate ii, dinitroplatinum; sulfuric acid, dihydrogen dinitrosulfatoplatinate ii solution, pt cont. ptShow More | |
platinum;sulfuric acid;dinitrite |
Specifications
12033-81-7 | |
Liquid | |
MFCD00064668 | |
Miscible with water. | |
[H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O | |
385.16 | |
384.17 (Anhydrous) | |
Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt) |
Brown | |
H2N2O8PtS | |
dihydrogen dinitrosulfatoplatinate ii, dinitroplatinum; sulfuric acid, dihydrogen dinitrosulfatoplatinate ii solution, pt cont. pt | |
SPPCQWREVYSZTJ-UHFFFAOYSA-N | |
platinum;sulfuric acid;dinitrite | |
102602532 | |
Brown liquid |
Safety and Handling
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H350
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.