missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99%, Thermo Scientific Chemicals

Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154

59.70 € - 133.67 €

Chemical Identifiers

CAS 108-18-9
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
PubChem CID 7912
IUPAC Name N-propan-2-ylpropan-2-amine
SMILES CC(C)NC(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10497160
View Documents
Thermo Scientific Acros
149460010
1 L
59.70 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10656922
View Documents
Thermo Scientific Acros
149460025
2.5 L
133.67 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-18-9
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
N-propan-2-ylpropan-2-amine
UAOMVDZJSHZZME-UHFFFAOYSA-N
7912
CC(C)NC(C)C

Specifications

108-18-9
Colorless
84.0°C
Authentic
Glass Bottle
[(CH3)2CH]2NH
1158
15, 3215
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
UAOMVDZJSHZZME-UHFFFAOYSA-N
N-propan-2-ylpropan-2-amine
7912
101.19
99%
Diisopropylamine, 99%
-61.0°C
0.7160g/mL
−17°C
99%
C6H15N
1.3910 to 1.3930
04, 154
0.716
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol, soluble in most common organic solvents
CC(C)NC(C)C
101.19
0.4 mPa.s (20°C)
95mbar at 20°C
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-558-5

Product Identifier
  • Diisopropylamine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only