missing translation for 'onlineSavingsMsg'
Learn More
Diisopropylamine, 99+%, Thermo Scientific Chemicals Product Code.: 10434335

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Product Code. 10434335
1 L, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10434335

Brand: Thermo Scientific Acros 219430010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008862
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym di-isopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, diisopropylamine, n-isopropylpropan-2-amine, n,n-diisopropylamine, dipa, diisopropyl amine, bis propan-2-yl amineShow MoreShow Less
PubChem CID 7912
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Density 0.7160 g/mL
Boiling Point 84.0°C
Flash Point -17°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C6H15N
Linear Formula [(CH3)2CH]2NH
Refractive Index 1.3910 to 1.3930
Quantity 1 L
Beilstein 04, 154
Merck Index 15, 3215
Specific Gravity 0.716
Solubility Information Solubility aq. soln.: 100 g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
IUPAC Name bis(propan-2-yl)amine
Viscosity 0.4 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diisopropylamine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469008 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number219432500,219430010,219430051
Lot NumberA0468284 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number219432500,219430010,219430051
Lot NumberA0465215 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number219432500,219430010,219430051
Lot NumberA0463740 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number219432500,219430010,219430051
Lot NumberA0462475 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number219432500,219430010,219430051
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diisopropylamine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.