Learn More
Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals
Dimethylamine is a used as a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber.
49.85 € - 182.91 €
Chemical Identifiers
CAS | 124-40-3 |
---|---|
Molecular Formula | C2H7N |
Molecular Weight (g/mol) | 45.085 |
MDL Number | MFCD00008288 |
InChI Key | ROSDSFDQCJNGOL-UHFFFAOYSA-N |
Synonym | dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution |
PubChem CID | 674 |
ChEBI | CHEBI:17170 |
IUPAC Name | N-methylmethanamine |
SMILES | CNC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369358
|
Thermo Scientific Alfa Aesar
043261.KV |
10 Ea. |
49.85 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379358
|
Thermo Scientific Alfa Aesar
043261.KW |
50 Ea. |
182.91 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dimethylamine is a used as a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrochemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-40-3 | |
45.085 | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
674 | |
N-methylmethanamine |
C2H7N | |
MFCD00008288 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
CHEBI:17170 | |
CNC |
Specifications
124-40-3 | |
15°C (60°F) | |
C2H7N | |
MFCD00008288 | |
605257 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.08 | |
Liquid |
0.89 | |
Ampoule | |
1.37 | |
UN1160 | |
0.89 | |
Fully miscible in water. | |
CNC | |
45.085 | |
CHEBI:17170 | |
40% w/w aq. soln. | |
Dimethylamine |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H314-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 204-697-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only