missing translation for 'onlineSavingsMsg'
Learn More
Dimethyltin dichloride, Thermo Scientific Chemicals Product Code.: 11364829

Dimethyltin dichloride, Thermo Scientific Chemicals

Product Code. 11364829
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11364829

Brand: Thermo Scientific Alfa Aesar 071142.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a pharmaceutical intermediate. It is employed as a coating, plastic and rubber auxilliary agent. Used as methyl Tin stabilizer intermediates, catalysts for esterification reaction, raw material for glass coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.68
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicityShow MoreShow Less
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Melting Point 103°C to 105°C
Boiling Point 185°C to 190°C
Quantity 2 g
UN Number UN3146
Beilstein 3660508
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water (20g/L (20°C)). Solubility in other solvents (0.1g/ml in methanol).
Formula Weight 219.69
Physical Form Crystalline
Chemical Name or Material Dimethyltin dichloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM29E018 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number071142.04,071142.18
Lot NumberW10K019 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number071142.04,071142.18
Lot NumberW11K023 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number071142.04,071142.18
Lot NumberY14I033 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog Number071142.04,071142.18
Lot NumberW19I023 Certificate TypeCertificate of Analysis Date24/10/2022 Catalog Number071142.04,071142.18
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethyltin dichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dimethyltin dichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyltin dichloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.