missing translation for 'onlineSavingsMsg'
Learn More
Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals Product Code.: 10178112

Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals

Product Code. 10178112
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10178112

Brand: Thermo Scientific Acros 409041000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26386-88-9
Molecular Formula C12H10N3O3P
Molecular Weight (g/mol) 275.20
MDL Number MFCD00001987
InChI Key SORGEQQSQGNZFI-UHFFFAOYSA-N
Synonym diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165avShow MoreShow Less
PubChem CID 123414
IUPAC Name [azido(phenoxy)phosphoryl]oxybenzene
SMILES [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Yellow to Amber
Density 1.2730g/mL
Boiling Point 157.0°C (0.2 mmHg)
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.5520
Quantity 100 g
Fieser 04,210; 05,280; 07,138; 08,211; 10,173
Merck Index 15,3366
Specific Gravity 1.273
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 275.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diphenylphosphoryl azide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457848 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number409040250,409045000,409041000
Lot NumberA0457848 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog Number409040250,409045000,409041000
Lot NumberA0445768 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number409040250,409045000,409041000
Lot NumberA0436900 Certificate TypeCertificate of Analysis Date30/11/2022 Catalog Number409040250,409045000,409041000
Lot NumberA0417773 Certificate TypeCertificate of Analysis Date23/03/2021 Catalog Number409040250,409045000,409041000
compliance-icons
Product Identifier
  • Diphenylphosphoryl azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.