Learn More
Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals
26.23 € - 203.01 €
Chemical Identifiers
CAS | 1135-99-5 |
---|---|
Molecular Formula | C12H12Cl2Sn |
Molecular Weight (g/mol) | 345.84 |
MDL Number | MFCD00000516 |
InChI Key | WSXPROXKZIMZIU-UHFFFAOYSA-N |
Synonym | diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr |
PubChem CID | 14342 |
IUPAC Name | dichloro(diphenyl)stannane |
SMILES | Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334829
|
Thermo Scientific Alfa Aesar
071139.04 |
2 g |
26.23 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314829
|
Thermo Scientific Alfa Aesar
071139.09 |
10 g |
63.42 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324829
|
Thermo Scientific Alfa Aesar
071139.18 |
50 g |
203.01 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in the manufacture of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1135-99-5 | |
345.84 | |
WSXPROXKZIMZIU-UHFFFAOYSA-N | |
14342 | |
Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1 |
C12H12Cl2Sn | |
MFCD00000516 | |
diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr | |
dichloro(diphenyl)stannane |
Specifications
1135-99-5 | |
White | |
>150°C (302°F) | |
(C6H5)2SnCl2 | |
Moisture sensitive | |
Sparingly Soluble (2.4E-3g/L) (25°C),Calc. | |
Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1 | |
345.84 | |
343.81 | |
Crystalline |
41°C to 43°C | |
333°C to 337°C (decomposition) | |
C12H12Cl2Sn | |
MFCD00000516 | |
diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr | |
WSXPROXKZIMZIU-UHFFFAOYSA-N | |
dichloro(diphenyl)stannane | |
14342 | |
95% | |
Diphenyltin dichloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 214-496-3
missing translation for 'rtecsNumber' : WH7253000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.