Learn More
Diphenylzinc, 98+%, Thermo Scientific Chemicals
154.00 € - 598.00 €
Chemical Identifiers
CAS | 1078-58-6 |
---|---|
Molecular Formula | C12H10Zn |
Molecular Weight (g/mol) | 219.59 |
MDL Number | MFCD00014071 |
InChI Key | BUNROVZNGITPIX-UHFFFAOYSA-N |
Synonym | diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2 |
PubChem CID | 517896 |
IUPAC Name | zinc;benzene |
SMILES | [Zn++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1 |
Descripción
Diphenylzinc is used as the synthetic equivalent of ph- synthon. It is also useful in a Michael 1,4-addition reactions. It is used as a catalyst for propylene oxide polymerization in benzene medium. Further, it is a reactive coupling compound in Negishi cross-coupling reactions. In addition to this, it utilized in the preparation of arylcopper, arylsilver and arylgold compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
1078-58-6 | |
219.59 | |
BUNROVZNGITPIX-UHFFFAOYSA-N | |
517896 | |
[Zn++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1 |
C12H10Zn | |
MFCD00014071 | |
diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2 | |
zinc;benzene |
Especificaciones
1078-58-6 | |
280°C to 285°C | |
MFCD00014071 | |
Air and moisture sensitive | |
Soluble in most organic solvents. | |
[Zn++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1 | |
219.59 | |
219.58 | |
Crystalline |
107°C | |
C12H10Zn | |
UN3181 | |
diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2 | |
BUNROVZNGITPIX-UHFFFAOYSA-N | |
zinc;benzene | |
517896 | |
≥98% | |
Diphenylzinc |
Seguridad y manipulación
GHS H Statement
H228
Flammable solid.
P210-P222-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H250-H314
missing translation for 'dotInformation' : Hazard Class: 4.2; Packaging Group: I
missing translation for 'einecsNumber' : 214-082-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.