Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand: Lovibond™ 230100
This item is not returnable.
View return policy
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
Iron | |
Comparator 2000+ | |
Color Comparator Disc |
Disc 3/116 | |
0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.7, 0.8, 1.0mg/L |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Lovibond™ Comparator Disc > Analyte: Iron
Spot an opportunity for improvement?Share a Content Correction