missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
178.00 € - 406.00 €
Product Code | Brand | Analytes | Description | Range | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Analytes | Description | Range | Price | Quantity & Availability | ||||||
13131681
|
Lovibond™
221130 |
ph Value | Disc 2/1J | 6.8, 7, 7.2, 7.4, 7.6, 7.8, 8, 8.2, 8.4 |
248.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13176030
|
Lovibond™
221080 |
ph Value | Disc 2/1E | 4.4, 4.6, 4.8, 5, 5.2, 5.4, 5.6, 5.8, 6 |
196.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13186030
|
Lovibond™
221110 |
ph Value | Disc 2/1H | 6, 6.2, 6.4, 6.6, 6.8, 7, 7.2, 7.4, 7.6 |
196.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13111681
|
Lovibond™
230100 |
Iron | Disc 3/116 | 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.7, 0.8, 1.0mg/L |
266.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13191671
|
Lovibond™
234010 |
Chlorine (DPD) | Disc 3/40A | 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.7, 0.8, 1.0mg/L |
248.00 €
Pack of 2000 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13126040
|
Lovibond™
233930 |
Chlorine | Disc 3/40K | 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6 mg/L |
266.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13101681
|
Lovibond™
234020 |
Chlorine | Disc 3/40B | 0.2, 0.4, 0.6, 1.0, 1.5, 2.0, 2.5, 3.0, 4.0mg/L |
266.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
13166030
|
Lovibond™
221030 |
ph Value | Disc 2/1B | 2.8, 3, 3.2, 3.4, 3.6, 3.8, 4, 4.2, 4.4 |
196.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13116040
|
Lovibond™
230070 |
Ammonia | Disc 3/113 | 0, 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.8, 1.0 mg/L N |
266.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13121681
|
Lovibond™
230360 |
Nitrate | Disc 3/142 | 10-100ppm N | N/A | N/A | N/A | |||||
13146040
|
Lovibond™
232110 |
Chlorine | Disc 3/2 Hypo | 2, 4, 6, 8, 10, 12, 14, 16% w/w available Cl2 |
406.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
13106040
|
Lovibond™
237000 |
Ortho-phosphate | Disc 3/70 | 0, 10, 20, 30, 40, 50, 60, 80, 100mg/L PO4 |
266.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
12744527
|
Lovibond™
283120 |
Ammonia | Disc NAB | 0.08, 0.24, 0.28, 0.32, 0.36, 0.4, 0.44, 0.48, 0.52mg/L NH3 |
266.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
12784527
|
Lovibond™
283680 |
Dissolved Oxygen | Disc NOE | 0, 0.005, 0.01, 0.015, 0.03, 0.055, 0.08, 0.1, 0.12 mL/L |
196.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
12794527
|
Lovibond™
283700 |
Hydrazine | Disc: NOH | 0, 0.5, 1, 2, 3, 4, 6, 8, 10ug |
266.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample – simply rotate disc for colour match
- Glass fade-free standards for longevity of use
- Shock resistant, scratch risistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consisist exclusively of through-and-through colored and flat-ground glasses
Specifications
Comparator 2000+ | |
Color Comparator Disc |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lovibond™ Comparator Disc